Assay Details
| Gene Symbol: BIRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048023.1 | NP_001041488.1 | 4 - 5 | 1066 | 88 | |
| XM_005619507.3 | XP_005619564.1 | 5 - 6 | 2483 | 88 | ||
| GenBank mRNA | DQ223014.1 | - | 4 - 5 | 1066 | 88 | |
More Information
Set Membership: |
|

BIRC2
Genomic Map