Assay Details
| Gene Symbol: EMR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038665.2 | NP_001033754.2 | 10 - 11 | 1206 | 72 | |
| XM_005632346.3 | XP_005632403.1 | 11 - 12 | 1337 | 72 | ||
| GenBank mRNA | DQ227278.1 | - | 10 - 11 | 1206 | 72 | |
More Information
Set Membership: |
|

EMR4
Genomic Map