Assay Details
| Gene Symbol: EMR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038665.2 | NP_001033754.2 | 3 - 4 | 214 | 99 | |
| XM_005632346.3 | XP_005632403.1 | 4 - 5 | 345 | 99 | ||
| GenBank mRNA | DQ227278.1 | - | 3 - 4 | 214 | 99 | |
More Information
Set Membership: |
|

EMR4
Genomic Map