Assay Details
| Gene Symbol: IGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313855.1 | NP_001300784.1 | 2 - 3 | 301 | 104 | |
| XM_014119233.2 | XP_013974708.1 | 2 - 3 | 257 | 104 | ||
| XM_022427670.1 | XP_022283378.1 | 2 - 3 | 483 | 104 | ||
| XM_861842.5 | XP_866935.1 | 2 - 3 | 255 | 104 | ||
| GenBank mRNA | L08254.1 | - | 1 - 2 | 131 | 104 | |
More Information
Set Membership: |
|

IGF1
Genomic Map