Assay Details
| Gene Symbol: SEC61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003326.1 | NP_001003326.1 | 3 - 4 | 306 | 84 | |
| GenBank mRNA | L25052.1 | - | 3 - 4 | 306 | 84 | |
More Information
Set Membership: |
|

SEC61B
Genomic Map