Assay Details
| Gene Symbol: PON2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003205.2 | NP_001003205.2 | 2 - 3 | 74 | ||
| GenBank mRNA | DN867252.1 | - | 2 - 3 | 74 | ||
| L48515.1 | - | 2 - 3 | 157 | 74 | ||
More Information
Set Membership: |
|

PON2
Genomic Map