Assay Details
| Gene Symbol: HLA-DRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014768.1 | NP_001014768.1 | 4 - 5 | 805 | 108 | |
| GenBank mRNA | M29611.1 | - | 4 - 5 | 805 | 108 | |
More Information
Set Membership: |
|

HLA-DRB1
Genomic Map