Assay Details
| Gene Symbol: DLA88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014767.1 | NP_001014767.1 | 2 - 3 | 340 | 67 | |
| GenBank mRNA | HQ340113.1 | - | 1 - 2 | 267 | 67 | |
| HQ340118.1 | - | 1 - 2 | 267 | 67 | ||
| JQ733514.1 | - | 1 - 2 | 271 | 67 | ||
| LC130507.1 | - | 1 - 2 | 267 | 67 | ||
| LC130508.1 | - | 1 - 2 | 267 | 67 | ||
| LC130512.1 | - | 1 - 2 | 267 | 67 | ||
| LC130515.1 | - | 1 - 2 | 267 | 67 | ||
| LC130516.1 | - | 1 - 2 | 267 | 67 | ||
| LC130518.1 | - | 1 - 2 | 267 | 67 | ||
| LC130520.1 | - | 1 - 2 | 267 | 67 | ||
| LC130522.1 | - | 1 - 2 | 267 | 67 | ||
| LC130523.1 | - | 1 - 2 | 267 | 67 | ||
| LC130525.1 | - | 1 - 2 | 267 | 67 | ||
| LC130526.1 | - | 1 - 2 | 267 | 67 | ||
| LC171423.1 | - | 1 - 2 | 267 | 67 | ||
| LC171424.1 | - | 1 - 2 | 267 | 67 | ||
| LC171425.1 | - | 1 - 2 | 267 | 67 | ||
| LC171429.1 | - | 1 - 2 | 267 | 67 | ||
| LC171431.1 | - | 1 - 2 | 267 | 67 | ||
| LC171432.1 | - | 1 - 2 | 267 | 67 | ||
| LC171434.1 | - | 1 - 2 | 267 | 67 | ||
| LC171436.1 | - | 1 - 2 | 267 | 67 | ||
| LC171437.1 | - | 1 - 2 | 267 | 67 | ||
| LC171438.1 | - | 1 - 2 | 267 | 67 | ||
| Gene Symbol: DLA-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | LC130546.1 | - | 2 - 3 | 331 | 67 | |
| Gene Symbol: LOC100685196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118307.2 | XP_013973782.1 | 2 - 3 | 426 | 67 | |
| GenBank mRNA | LC130521.1 | - | 1 - 2 | 267 | 67 | |
More Information
Set Membership: |
|

DLA88
Genomic Map