Assay Details
| Gene Symbol: RAB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003316.1 | NP_001003316.1 | 539 | 75 | ||
| XM_005632015.2 | XP_005632072.1 | 758 | 75 | |||
| GenBank mRNA | M35522.1 | - | 539 | 75 | ||
More Information
Set Membership: |
|

RAB7A
Genomic Map