Assay Details
| Gene Symbol: DIO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007126.2 | NP_001007127.1 | 3 - 4 | 693 | 85 | |
| GenBank mRNA | U11762.1 | - | 3 - 4 | 701 | 85 | |
More Information
Set Membership: |
|

DIO1
Genomic Map