Assay Details
| Gene Symbol: SPCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003324.2 | NP_001003324.1 | 1 - 2 | 137 | 126 | |
| GenBank mRNA | U12687.1 | - | 1 - 2 | 138 | 126 | |
More Information
Set Membership: |
|

SPCS2
Genomic Map