Assay Details
| Gene Symbol: PRPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003289.1 | NP_001003289.1 | 1 - 2 | 827 | 70 | |
| GenBank mRNA | U27349.1 | - | 1 - 2 | 827 | 70 | |
| U36577.1 | - | 1 - 2 | 587 | 70 | ||
| X94122.1 | - | 763 | 70 | |||
More Information
Set Membership: |
|

PRPH2
Genomic Map