Assay Details
| Gene Symbol: PLA2G7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003198.1 | NP_001003198.1 | 9 - 10 | 961 | 67 | |
| XM_005627023.3 | XP_005627080.1 | 9 - 10 | 1119 | 67 | ||
| XM_005627024.3 | XP_005627081.1 | 9 - 10 | 1058 | 67 | ||
| XM_005627025.3 | XP_005627082.1 | 9 - 10 | 1036 | 67 | ||
| XM_005627026.3 | XP_005627083.1 | 9 - 10 | 920 | 67 | ||
| XM_022425715.1 | XP_022281423.1 | 9 - 10 | 1029 | 67 | ||
| GenBank mRNA | U34246.1 | - | 9 - 10 | 961 | 67 | |
More Information
Set Membership: |
|

PLA2G7
Genomic Map