Assay Details
| Gene Symbol: PDE6G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003235.1 | NP_001003235.1 | 358 | 62 | ||
| XM_022422536.1 | XP_022278244.1 | 1080 | 62 | |||
| GenBank mRNA | U49359.1 | - | 358 | 62 | ||
More Information
Set Membership: |
|

PDE6G
Genomic Map