Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003251.1 | NP_001003251.1 | 2 - 3 | 99 | 77 | |
| GenBank mRNA | U57440.1 | - | 2 - 3 | 99 | 77 | |
More Information
Set Membership: |
|

SRP14
Genomic Map