Assay Details
| Gene Symbol: SRP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003272.1 | NP_001003272.1 | 12 - 13 | 1343 | 78 | |
| XM_005623180.3 | XP_005623237.1 | 11 - 12 | 1543 | 78 | ||
| XM_005623181.3 | XP_005623238.1 | 11 - 12 | 1149 | 78 | ||
| GenBank mRNA | X16318.1 | - | 1343 | 78 | ||
More Information
Set Membership: |
|

SRP54
Genomic Map