Assay Details
| Gene Symbol: ANXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003255.1 | NP_001003255.1 | 232 | 116 | ||
| XM_005627805.3 | XP_005627862.1 | 166 | 116 | |||
| XM_005627806.2 | XP_005627863.1 | 205 | 116 | |||
| GenBank mRNA | X80208.1 | - | 149 | 116 | ||
| X80209.1 | - | 232 | 116 | |||
More Information
Set Membership: |
|

ANXA13
Genomic Map