Assay Details
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003253.1 | NP_001003253.1 | 1 - 2 | 117 | 66 | |
| GenBank mRNA | X92505.1 | - | 117 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003253.1 | NP_001003253.1 | 1 - 2 | 117 | 66 | |
| GenBank mRNA | X92505.1 | - | 117 | 66 | ||
Set Membership: |
|