Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003218.1 | NP_001003218.1 | 265 | 93 | ||
| XM_005630068.3 | XP_005630125.1 | 1422 | 93 | |||
| XM_022404319.1 | XP_022260027.1 | 785 | 93 | |||
| GenBank mRNA | X99144.1 | - | 265 | 93 | ||
More Information
Set Membership: |
|

TRIB2
Genomic Map