Assay Details
| Gene Symbol: KLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003284.1 | NP_001003284.1 | 3 - 4 | 522 | 92 | |
| GenBank mRNA | Y00751.1 | - | 522 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003284.1 | NP_001003284.1 | 3 - 4 | 522 | 92 | |
| GenBank mRNA | Y00751.1 | - | 522 | 92 | ||
Set Membership: |
|