Assay Details
| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003177.1 | NP_001003177.1 | 1 - 2 | 58 | 71 | |
| XM_005640983.3 | XP_005641040.1 | 2 - 3 | 166 | 71 | ||
| XM_022415491.1 | XP_022271199.1 | 2 - 3 | 455 | 71 | ||
| GenBank mRNA | Y18136.2 | - | 58 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003177.1, XM_005640983.3, XM_022415491.1, Y18136.2 transcripts.
Set Membership: |
|

AVPR2
Genomic Map