Assay Details
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421726.1 | XP_022277434.1 | 5 - 6 | 568 | 88 | |
| GenBank mRNA | Z22818.1 | - | 400 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421726.1 | XP_022277434.1 | 5 - 6 | 568 | 88 | |
| GenBank mRNA | Z22818.1 | - | 400 | 88 | ||
Set Membership: |
|