Assay Details
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003208.1 | NP_001003208.1 | 3 - 4 | 289 | 69 | |
| GenBank mRNA | Z22820.1 | - | 289 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003208.1 | NP_001003208.1 | 3 - 4 | 289 | 69 | |
| GenBank mRNA | Z22820.1 | - | 289 | 69 | ||
Set Membership: |
|