Assay Details
| Gene Symbol: BIRC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003348.1 | NP_001003348.1 | 405 | 140 | ||
| XM_022422541.1 | XP_022278249.1 | 1596 | 140 | |||
| GenBank mRNA | AB180206.1 | - | 405 | 140 | ||
| AY741504.1 | - | 389 | 140 | |||
| KT693114.1 | - | 383 | 140 | |||
More Information
Set Membership: |
|

BIRC5
Genomic Map