Assay Details
| Gene Symbol: MADCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024639.1 | NP_001019810.1 | 1 - 2 | 81 | 139 | |
| GenBank mRNA | AB211528.1 | - | 1 - 2 | 81 | 139 | |
More Information
Set Membership: |
|

MADCAM1
Genomic Map