Assay Details
| Gene Symbol: CTSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195834.1 | NP_001182763.1 | 1 - 2 | 381 | 148 | |
| GenBank mRNA | AF060171.1 | - | 1 - 2 | 247 | 148 | |
| GR899188.1 | - | 381 | 148 | |||
More Information
Set Membership: |
|

CTSC
Genomic Map