Assay Details
| Gene Symbol: RPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003099.1 | NP_001003099.1 | 1 - 2 | 162 | 87 | |
| GenBank mRNA | AJ388523.1 | - | 1 - 2 | 162 | 87 | |
More Information
Set Membership: |
|

RPS17
Genomic Map