Assay Details
| Gene Symbol: RPL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286939.1 | NP_001273868.1 | 2 - 3 | 182 | 83 | |
| GenBank mRNA | AJ388527.1 | - | 1 - 2 | 127 | 83 | |
| DN318367.1 | - | 1 - 2 | 132 | 83 | ||
| DN353284.1 | - | 2 - 3 | 183 | 83 | ||
| GR889306.1 | - | 2 - 3 | 152 | 83 | ||
More Information
Set Membership: |
|

RPL7A
Genomic Map