Assay Details
| Gene Symbol: BCL2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031635.2 | NP_001026805.1 | 3 - 4 | 605 | 131 | |
| XM_005623202.3 | XP_005623259.1 | 3 - 4 | 648 | 131 | ||
| GenBank mRNA | DQ116955.1 | - | 1 - 2 | 439 | 131 | |
More Information
Set Membership: |
|

BCL2L2
Genomic Map