Assay Details
| Gene Symbol: COX5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144130.1 | NP_001137602.1 | 2 - 3 | 172 | 77 | |
| XM_005625686.3 | XP_005625743.1 | 2 - 3 | 313 | 77 | ||
| XM_005625687.3 | XP_005625744.1 | 2 - 3 | 243 | 77 | ||
| GenBank mRNA | AM260640.1 | - | 2 - 3 | 172 | 77 | |
More Information
Set Membership: |
|

COX5B
Genomic Map