Assay Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406449.1 | XP_022262157.1 | 1053 | 130 | ||
XM_022406450.1 | XP_022262158.1 | 934 | 130 | |||
XM_022406451.1 | XP_022262159.1 | 952 | 130 | |||
XM_022406452.1 | XP_022262160.1 | 754 | 130 | |||
XM_533762.7 | XP_533762.3 | 1065 | 130 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
