Assay Details
| Gene Symbol: C7H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421373.1 | XP_022277081.1 | 5 - 6 | 809 | 69 | |
| XM_022421375.1 | XP_022277083.1 | 6 - 7 | 895 | 69 | ||
| XM_022421376.1 | XP_022277084.1 | 5 - 6 | 790 | 69 | ||
| XM_847193.4 | XP_852286.2 | 6 - 7 | 903 | 69 | ||
More Information
Set Membership: |
|

C7H1orf43
Genomic Map