Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145150.1 | NP_001138622.1 | 10 - 11 | 1563 | 83 | |
| XM_005618880.3 | XP_005618937.1 | 11 - 12 | 1721 | 83 | ||
More Information
Set Membership: |
|

BMPR1A
Genomic Map