Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632662.2 | XP_005632719.1 | 3 - 4 | 478 | 81 | |
| XM_005632663.2 | XP_005632720.1 | 3 - 4 | 473 | 81 | ||
| XM_005632664.2 | XP_005632721.1 | 3 - 4 | 472 | 81 | ||
| XM_022406829.1 | XP_022262537.1 | 2 - 3 | 431 | 81 | ||
More Information
Set Membership: |
|

RFXANK
Genomic Map