Assay Details
| Gene Symbol: LOC100685226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427692.1 | XP_022283400.1 | 3 - 4 | 477 | 74 | |
| XM_022427695.1 | XP_022283403.1 | 3 - 4 | 493 | 74 | ||
| XM_022427697.1 | XP_022283405.1 | 1 - 2 | 125 | 74 | ||
| XM_022427707.1 | XP_022283415.1 | 3 - 4 | 477 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022427695.1, XM_022427697.1 transcripts.
Set Membership: |
|

LOC100685226
Genomic Map