Assay Details
| Gene Symbol: LOC100685226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427707.1 | XP_022283415.1 | 4 - 5 | 570 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100685226| Gene Symbol: LOC100685226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427707.1 | XP_022283415.1 | 4 - 5 | 570 | 113 | |
Set Membership: |
|