Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629145.3 | XP_005629202.1 | 3 - 4 | 504 | 93 | |
| XM_005629146.3 | XP_005629203.1 | 3 - 4 | 842 | 93 | ||
| XM_005629149.2 | XP_005629206.1 | 2 - 3 | 264 | 93 | ||
| XM_022428092.1 | XP_022283800.1 | 2 - 3 | 385 | 93 | ||
| XM_022428093.1 | XP_022283801.1 | 2 - 3 | 388 | 93 | ||
| XM_022428094.1 | XP_022283802.1 | 2 - 3 | 387 | 93 | ||
| XM_022428095.1 | XP_022283803.1 | 2 - 3 | 387 | 93 | ||
More Information
Set Membership: |
|

TPPP2
Genomic Map