Assay Details
| Gene Symbol: SIAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617594.3 | XP_005617651.1 | 1 - 2 | 139 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005617594.3 transcripts.
Set Membership: |
|

SIAH1
Genomic Map