Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621739.3 | XP_005621796.1 | 1 - 2 | 156 | 67 | |
| XM_547207.5 | XP_547207.2 | 1 - 2 | 156 | 67 | ||
More Information
Set Membership: |
|

CIAO3
Genomic Map