Assay Details
| Gene Symbol: LOC100856034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639595.4 | XP_003639643.1 | 7 - 8 | 1041 | 90 | |
| XM_005630401.3 | XP_005630458.1 | 8 - 9 | 1151 | 90 | ||
| XM_014120407.2 | XP_013975882.1 | 7 - 8 | 1062 | 90 | ||
| XM_022404699.1 | XP_022260407.1 | 8 - 9 | 1147 | 90 | ||
More Information
Set Membership: |
|

LOC100856034
Genomic Map