Assay Details
| Gene Symbol: NT5C3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628704.3 | XP_005628761.1 | 8 - 9 | 698 | 111 | |
| XM_005628705.3 | XP_005628762.1 | 8 - 9 | 743 | 111 | ||
| XM_022427558.1 | XP_022283266.1 | 7 - 8 | 636 | 111 | ||
| XM_022427559.1 | XP_022283267.1 | 7 - 8 | 615 | 111 | ||
| XM_847048.4 | XP_852141.1 | 7 - 8 | 790 | 111 | ||
More Information
Set Membership: |
|

NT5C3A
Genomic Map