Assay Details
| Gene Symbol: CRIPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117513.2 | XP_013972988.1 | 3 - 4 | 298 | 140 | |
| XM_846811.5 | XP_851904.1 | 4 - 5 | 353 | 140 | ||
More Information
Set Membership: |
|

CRIPT
Genomic Map