Assay Details
Gene Symbol: ARL5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846551.5 | XP_851644.1 | 894 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
