Assay Details
| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620142.3 | XP_005620199.1 | 2 - 3 | 239 | 62 | |
| XM_005620143.3 | XP_005620200.1 | 2 - 3 | 254 | 62 | ||
| XM_846726.5 | XP_851819.2 | 2 - 3 | 276 | 62 | ||
More Information
Set Membership: |
|

SHMT1
Genomic Map