Assay Details
| Gene Symbol: ZNF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419234.1 | XP_022274942.1 | 2 - 3 | 145 | 69 | |
| XM_022419235.1 | XP_022274943.1 | 2 - 3 | 145 | 69 | ||
| XM_022419236.1 | XP_022274944.1 | 4 - 5 | 654 | 69 | ||
| XM_022419237.1 | XP_022274945.1 | 5 - 6 | 411 | 69 | ||
| XM_022419238.1 | XP_022274946.1 | 4 - 5 | 704 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022419234.1, XM_022419235.1, XM_022419238.1 transcripts.
Set Membership: |
|

ZNF23
Genomic Map