Assay Details
| Gene Symbol: AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617613.2 | XP_005617670.1 | 5 - 6 | 595 | 62 | |
| XM_014109721.2 | XP_013965196.1 | 5 - 6 | 466 | 62 | ||
| XM_858927.5 | XP_864020.1 | 5 - 6 | 594 | 62 | ||
More Information
Set Membership: |
|

AK2
Genomic Map