Assay Details
| Gene Symbol: TAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287043.1 | NP_001273972.1 | 5 - 6 | 640 | 84 | |
| XM_005617698.3 | XP_005617755.1 | 6 - 7 | 740 | 84 | ||
| GenBank mRNA | DN756363.1 | - | 1 | 249 | 84 | |
| DR105471.1 | - | 5 - 6 | 653 | 84 | ||
More Information
Set Membership: |
|

TAF12
Genomic Map