Assay Details
| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253744.1 | NP_001240673.1 | 628 | 78 | ||
| GenBank mRNA | DN872200.1 | - | 404 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253744.1 | NP_001240673.1 | 628 | 78 | ||
| GenBank mRNA | DN872200.1 | - | 404 | 78 | ||