Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434038.4 | XP_003434086.1 | 4 - 5 | 666 | 65 | |
| XM_003434039.4 | XP_003434087.1 | 3 - 4 | 526 | 65 | ||
| XM_846277.5 | XP_851370.1 | 2 - 3 | 849 | 65 | ||
More Information
Set Membership: |
|

PITX2
Genomic Map