Assay Details
| Gene Symbol: METTL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122786.2 | XP_013978261.1 | 4 - 5 | 847 | 53 | |
| XM_534100.6 | XP_534100.3 | 4 - 5 | 1124 | 53 | ||
More Information
Set Membership: |
|

METTL15
Genomic Map